siRNA Details
Gene Symbol: Gm15201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136312.1 | - | 2 | 382 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB624209.1 | - | 2 | 378 | Chr. X - 162735639 - 162736611 [-] on Build GRCm38 |
