siRNA Details
Gene Symbol: 4933430H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045855.1 | 2 | 270 | |||
GenBank mRNA | AK016994.1 | - | 2 | 271 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716678.1 | - | 2 | 271 | Chr. 7 - 82939680 - 82971391 [-] on Build GRCm38 |
