siRNA Details
Gene Symbol: 2810428J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018041.1 | - | 1 | 27 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717971.1 | - | 1 | 25 | Chr. 3 - 135427075 - 135427930 [-] on Build GRCm38 |
