siRNA Details
Gene Symbol: AW495222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045086.1 | 2, 3 | 527 | |||
GenBank mRNA | AK046172.1 | - | 2, 3 | 528 | ||
BB314344.2 | - | 2, 3 | 350 | |||
BB644732.1 | - | 2, 3 | 525 | |||
BE656108.1 | - | 2, 3 | 410 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712700.1 | - | 2, 3 | 499 | Chr. 13 - 93706282 - 93719146 [-] on Build GRCm38 |
