siRNA Details
Gene Symbol: AW495222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045086.1 | 2 | 478 | |||
GenBank mRNA | AK046172.1 | - | 2 | 479 | ||
BB314344.2 | - | 2 | 301 | |||
BB644732.1 | - | 2 | 476 | |||
BE656108.1 | - | 2 | 361 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712700.1 | - | 2 | 450 | Chr. 13 - 93706282 - 93719146 [-] on Build GRCm38 |
