siRNA Details
Gene Symbol: 4930562F07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038030.1 | 3 | 653 | |||
NR_038031.1 | 3 | 581 | ||||
GenBank mRNA | AK016191.1 | - | 3 | 654 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA466557.1 | - | 3 | 577 | Chr. 1 - 160074822 - 160077977 [-] on Build GRCm38 |
