siRNA Details
Gene Symbol: 2310030A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040603.1 | 4 | 573 | |||
GenBank mRNA | AK009534.1 | - | 4 | 574 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709227.1 | - | 4 | 574 | Chr. 1 - 150320681 - 150346771 [-] on Build GRCm38 |
