siRNA Details
Gene Symbol: Gm33586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042407.1 | - | 1 | 75 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF537429.1 | - | 1 | 75 | Chr. 7 - 113009114 - 113010892 [+] on Build GRCm38 |
