siRNA Details
Gene Symbol: B830042I05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021034.1 | - | 2 | 467 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727810.1 | - | 2 | 467 | Chr. 8 - 11075895 - 11076638 [+] on Build GRCm38 |
