siRNA Details
Gene Symbol: C030002A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021037.1 | - | 1 | 191 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728097.1 | - | 1 | 191 | Chr. 8 - 17438796 - 17439150 [-] on Build GRCm38 |
