siRNA Details
Gene Symbol: 5430400D12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017243.1 | - | 2 | 466 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717054.1 | - | 2 | 463 | Chr. 7 - 79920757 - 79921500 [+] on Build GRCm38 |
