siRNA Details
Gene Symbol: 1700057H21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006824.1 | - | 1 | 27 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706809.1 | - | 1 | 27 | Chr. 2 - 51421050 - 51437900 [-] on Build GRCm38 |
