siRNA Details
Gene Symbol: 1700057A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006821.1 | - | 1 | 373 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706803.1 | - | 1 | 371 | Chr. 2 - 3148803 - 3149231 [+] on Build GRCm38 |
