siRNA Details
Gene Symbol: A930033C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020921.1 | - | 4 | 579 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742315.1 | - | 4 | 579 | Chr. 5 - 35397372 - 35400019 [-] on Build GRCm38 |
