siRNA Details
Gene Symbol: Gm38554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC049722.1 | - | 3 | 406 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU962724.1 | - | 3 | 409 | Chr. 5 - 119959243 - 119963370 [+] on Build GRCm38 |
