siRNA Details
Gene Symbol: A930033M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020926.1 | - | 1 | 336 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742326.1 | - | 1 | 336 | Chr. 6 - 40470590 - 40471280 [-] on Build GRCm38 |
