siRNA Details
Gene Symbol: Gm35156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156776.1 | - | 2 | 83 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY751640.1 | - | 2 | 83 | Chr. X - 17537950 - 17551333 [+] on Build GRCm38 |
