siRNA Details
Gene Symbol: Gm31048 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030263.1 | - | 3 | 755 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV283834.2 | - | 2 | 413 | Chr. 5 - 104239313 - 104243450 [-] on Build GRCm38 |
