siRNA Details
Gene Symbol: 3110035F07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014124.1 | - | 1 | 206 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713109.1 | - | 1 | 206 | Chr. 14 - 117665575 - 117666031 [+] on Build GRCm38 |
