siRNA Details
Gene Symbol: 3110035G12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014125.1 | - | 1 | 660 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713110.1 | - | 1 | 651 | Chr. 6 - 56776417 - 56777339 [-] on Build GRCm38 |
