siRNA Details
Gene Symbol: Gm26627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084376.1 | - | 2 | 474 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB658168.1 | - | 2 | 474 | Chr. 6 - 29507725 - 29508864 [+] on Build GRCm38 |
