siRNA Details
Gene Symbol: A230005M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038413.1 | - | 1 | 654 | ||
AK046328.1 | - | 1 | 649 | |||
AK082064.1 | - | 2 | 805 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A230005M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038413.1 | - | 1 | 654 | ||
AK046328.1 | - | 1 | 649 | |||
AK082064.1 | - | 2 | 805 |