siRNA Details
Gene Symbol: Irx3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052999.1 | - | 1 | 572 |
Gene Symbol: LOC101055810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC100534.1 | - | 3 | 1118 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB522863.2 | - | 1 | 450 | Chr. 8 - 91804563 - 91805255 [+] on Build GRCm38 |
