siRNA Details
Gene Symbol: Panct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013716.1 | - | 1 | 1060 | ||
AK156552.1 | - | 1 | 3443 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV472318.1 | - | 1 | 328 | Chr. X - 159537600 - 159538091 [+] on Build GRCm38 |
