siRNA Details
Gene Symbol: Panct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013716.1 | - | 1 | 1493 | ||
AK156552.1 | - | 1 | 3876 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Panct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013716.1 | - | 1 | 1493 | ||
AK156552.1 | - | 1 | 3876 |