siRNA Details
Gene Symbol: 4930555M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019767.1 | - | 2 | 261 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715739.1 | - | 2 | 261 | Chr. 3 - 5512993 - 5515333 [-] on Build GRCm38 |
