siRNA Details
Gene Symbol: 4930552P06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019761.1 | - | 1 | 244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715702.1 | - | 1 | 244 | Chr. 4 - 107596780 - 107597226 [-] on Build GRCm38 |
