siRNA Details
Gene Symbol: Gm30329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045708.1 | - | 1 | 123 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB644231.1 | - | 1 | 123 | Chr. 8 - 79225930 - 79230801 [+] on Build GRCm38 |
