siRNA Details
Gene Symbol: A030001D20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003910.1 | - | 2 | 155 | ||
AK037147.1 | - | 2 | 171 | |||
AK049852.1 | - | 2 | 157 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A030001D20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003910.1 | - | 2 | 155 | ||
AK037147.1 | - | 2 | 171 | |||
AK049852.1 | - | 2 | 157 |