siRNA Details
Gene Symbol: Gm31718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050164.1 | - | 4 | 298 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB653745.1 | - | 4 | 298 | Chr. 8 - 126820193 - 126876438 [-] on Build GRCm38 |
