siRNA Details
Gene Symbol: 1700010J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040579.1 | 1 | 44 | |||
GenBank mRNA | AK005834.1 | - | 1 | 44 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU938461.1 | - | 85 | Chr. - - [-] on Build |
