siRNA Details
Gene Symbol: 1700010K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005839.1 | - | 1, 2 | 104 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705987.1 | - | 1, 2 | 104 | Chr. 3 - 108248909 - 108256985 [-] on Build GRCm38 |
