siRNA Details
Gene Symbol: 1500035N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005363.1 | - | 2 | 778 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705531.1 | - | 2 | 778 | Chr. 5 - 24985842 - 24998168 [+] on Build GRCm38 |
