siRNA Details
Gene Symbol: Gm35915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078274.1 | - | 1 | 775 | ||
AK148359.1 | - | 1 | 794 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV232097.2 | - | 1 | 508 | Chr. 9 - 95407789 - 95408400 [+] on Build GRCm38 |
