siRNA Details
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017658.1 | - | 1 | 251 | ||
AK141302.1 | - | 1 | 1545 | |||
BC033603.1 | - | 1 | 2943 |

More Information
Other Species Targeted

