siRNA Details
Gene Symbol: 9530014B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020556.1 | - | 3 | 730 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV061028.1 | - | 3 | 769 | Chr. 13 - 53635894 - 53639191 [-] on Build GRCm38 |
