siRNA Details
Gene Symbol: A430106A12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079275.1 | - | 1 | 1216 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX633015.1 | - | 1 | 411 | Chr. 11 - 61879095 - 61879669 [-] on Build GRCm38 |
