siRNA Details
Gene Symbol: LOC102636700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134928.1 | - | 2 | 1243 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD363411.1 | - | 2 | 653 | Chr. 9 - 75310416 - 75314281 [-] on Build GRCm38 |
