siRNA Details
Gene Symbol: C030009J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044617.1 | - | 1 | 2069 | ||
BB283503.1 | - | 1 | 482 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB280653.2 | - | 1 | 646 | Chr. 12 - 102271812 - 102272487 [+] on Build GRCm38 |
