siRNA Details
Gene Symbol: Gm26698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087306.1 | - | 2 | 530 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664513.1 | - | 2 | 530 | Chr. 12 - 86891756 - 86892567 [-] on Build GRCm38 |
