siRNA Details
Gene Symbol: 1700019H22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006123.1 | - | 1 | 362 | ||
AK145683.1 | - | 1 | 345 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706229.1 | - | 1 | 362 | Chr. 8 - 80859386 - 80860017 [+] on Build GRCm38 |
