siRNA Details
Gene Symbol: Gm17182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087045.1 | - | 1 | 296 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664013.1 | - | 1 | 296 | Chr. 5 - 53047924 - 53049187 [-] on Build GRCm38 |
