siRNA Details
Gene Symbol: 2310002F09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077063.1 | 3 | 843 | |||
GenBank mRNA | AK009091.1 | - | 3 | 844 | ||
AK086402.1 | - | 1 | 283 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310002F09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077063.1 | 3 | 843 | |||
GenBank mRNA | AK009091.1 | - | 3 | 844 | ||
AK086402.1 | - | 1 | 283 |