siRNA Details
Gene Symbol: Gm10619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077222.1 | 1 | 555 | |||
GenBank mRNA | AK030117.1 | - | 309 | |||
AK133252.1 | - | 1 | 558 | |||
DQ543357.1 | - | 1 | 18 | |||
DQ548756.1 | - | 1 | 15 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB616852.1 | - | 2 | 309 | Chr. 7 - 73784069 - 73816431 [-] on Build GRCm38 |
