siRNA Details
Gene Symbol: Lemd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032278.1 | - | 1 | 649 | ||
BC066109.1 | - | 2 | 1767 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lemd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032278.1 | - | 1 | 649 | ||
BC066109.1 | - | 2 | 1767 |