siRNA Details
Gene Symbol: Lemd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032278.1 | - | 1 | 4049 | ||
BC066109.1 | - | 2 | 5167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE862032.1 | - | 1 | 415 | Chr. 1 - 132209028 - 132209503 [+] on Build GRCm38 |
