siRNA Details
Gene Symbol: 4930407I02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019572.1 | - | 2 | 155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714649.1 | - | 2 | 155 | Chr. 6 - 143232337 - 143233356 [-] on Build GRCm38 |
