siRNA Details
Gene Symbol: 4930420G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019579.1 | - | 1 | 34 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714717.1 | - | 1 | 34 | Chr. 16 - 88426374 - 88427066 [+] on Build GRCm38 |
