siRNA Details
Gene Symbol: B930092H01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045334.1 | 3 | 171 | |||
GenBank mRNA | AK047575.1 | - | 3 | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB647450.1 | - | 3 | 169 | Chr. 9 - 61261325 - 61293807 [-] on Build GRCm38 |
