siRNA Details
Gene Symbol: Gm31592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034176.1 | - | 3 | 490 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB626137.1 | - | 3 | 488 | Chr. 10 - 91888830 - 91909534 [+] on Build GRCm38 |
